missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Mark-It™ Tissue Marking Dyes
For a variety of tissue identification purposes use Epredia™ Mark-It™ Tissue Marking Dyes.
Brand: Epredia™ 3120125
64.28 GBP valid until 2024-10-31
Use promo code "21628" to get your promotional price.
This item is not returnable.
View return policy
Description
Features:- Permanent dyes that maintain their original brilliance during routine tissue processing cycles
- Can be used on fresh or formalin-fixed tissues
Uses include:
- Marking surgical margins
- Identifying serial segments of large tissues to ensure proper orientation and embedding
- Maintaining proper tissue orientation for Mohs surgery specimens
- Identification of multiple pieces of tissues processed in the same cassette
Ordering Information:
- Available in black, blue, green, orange, red and yellow in 2 oz. bottles
Specifications
Black | |
Tissue Marking Dye | |
2 oz./ ea. |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Danger
Hazard Category
- Carcinogenicity Category 2
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H351-Suspected of causing cancer.
- H371-May cause damage to organs.
Precautionary Statement
- P362-Take off contaminated clothing.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash face, hands, and anyexposed skin thoroughly after handling.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P271-Use only outdoors or in a well-ventilated area.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P405-Store locked up.
Supplemental information
- MIXTURE LIST-Contains : carbon black, ammonium hydroxide, formaldehyde