missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Neg-50™ Frozen Section Medium
Yield high-quality sections down to -50°C without chatter using water-soluble Epredia™ Neg-50™ Frozen Section Medium. Comes in clear and five additional colors with all leaving no residue after staining.
£39.50 - £142.00
Product Code | Brand | Color | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Quantity | Price | Quantity & Availability | ||||
11912365
|
Epredia™
6502 |
Clear | 4 oz. per bottle |
£63.20
Pack of 2 |
In Stock
Log in to see stock. |
||||
12678086
|
Epredia™
6502B |
Blue | 4 oz.per bottle |
£65.90
Pack of 2 |
Estimated Shipment: 10-07-2024
Log in to see stock. |
||||
12688086
|
Epredia™
6502G |
Green | 4 oz. per bottle |
£66.90
Pack of 2 |
Estimated Shipment: 10-07-2024
Log in to see stock. |
||||
12698086
|
Epredia™
6502O |
Orange | 4 oz. per bottle |
£49.80
Pack of 2 |
Estimated Shipment: 31-07-2024
Log in to see stock. |
||||
12618096
|
Epredia™
6502Y |
Yellow | 4 oz. per bottle |
£55.95
Pack of 2 |
Estimated Shipment: 31-07-2024
Log in to see stock. |
||||
12647756
|
Epredia™
6506 |
Clear | 4 oz. per bottle |
£91.55
Pack of 6 |
N/A | ||||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
12608096
|
Epredia™
6502P |
Pink | 4 oz. per bottle |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12628096
|
Epredia™
6506M |
Five Colors plus Clear | 4 oz. per bottle |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Yield high-quality sections down to -50°C without chatter using Epredia; Neg-50™ Frozen Section Medium, a water-soluble frozen section medium.
- Buffered to ensure proper alkalinity (pH = 8.0); leaves no residue after staining
- Specially formulated to prevent tissue separation from the slide
Ordering Information:
Available in five colors for easy identification of specimens: pink, blue, green, yellow, and orange. Multi-packs contain one of each color.