missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Richard-Allan Scientific™ Periodic Acid-Schiff (PAS), Schiff Reagent

Demonstrate the patterns and presence of specific carbohydrates with Epredia™ Richard-Allan Scientific™ Periodic Acid-Schiff (PAS) Solution

Brand:  Epredia™ 88018

Code : A0

Additional Details : Weight : 0.01000kg


 View more versions of this product

Product Code. 12513647

  • £73.55 / 500mL
Estimated Shipment: 26-09-2024
to see stock.
Add to basket

Description

Description

  • Glycogen, mucin, fungal walls and some basement membranes stain rose to magenta
  • Nuclei stain blue
  • Kit includes: Hematoxylin I, periodic acid solution (0.5%), and Schiff reagent
Specifications

Specifications

Hematoxylin I
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Hematoxylin 1

Signal Word
  • Warning

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P370+P378-In case of fire: Use water/water spray/water jet/carbon dioxide/sand/foam/alcoholresistant-foam/chemical resistant powde to extinguish.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : nitrocellulose

SDS
Documents

Documents

Certificates
Special Offers

Special Offers