missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Richard-Allan Scientific™ Periodic Acid-Schiff (PAS), Schiff Reagent

Demonstrate the patterns and presence of specific carbohydrates with Epredia™ Richard-Allan Scientific™ Periodic Acid-Schiff (PAS) Solution

Brand:  Epredia™ 87007

Code : LE

Additional Details : Weight : 2.07176kg


 View more versions of this product

Product Code. 12562696

  • £184.00 / Each
Estimated Shipment: 07-11-2024
to see stock.
Add to basket

Description

Description

  • Glycogen, mucin, fungal walls and some basement membranes stain rose to magenta
  • Nuclei stain blue
  • Kit includes: Hematoxylin I, periodic acid solution (0.5%), and Schiff reagent
Specifications

Specifications

Periodic Acid Schiff
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H371-May cause damage to organs.

Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P234-Keep only in original container.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P361 + P364-Take off immediately all contaminated clothing and wash it before reuse.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P390-Absorb spillage to prevent material damage.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P406-Store in corrosive resistant/polypropylene container with a resistant inner liner.
  • P402-Store in a dry place.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : hydrochloric acid, potassium ferrocyanide, Basic Fuchsin, high purity biologicalstain, special for

SDS
Documents

Documents

Certificates
Special Offers

Special Offers