missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Resolve™ Immersion Oil

Eliminate toxicity and disposal problems with the Epredia™ Resolve™ Immersion Oil.

Brand:  Epredia™ M2004

Code : A0

Additional Details : Weight : 0.01000kg


 View more versions of this product

Product Code. 12550037

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Manufactured using a proprietary process that removes interfering substances and will not damage valuable optics or irreplaceable slide specimens
  • Packaged conveniently in four sizes
Product Details:
  • Available in both low and high viscosity
  • Non-drying and non-hardening
  • Refractive index of 1.5150
Specifications

Specifications

Immersion Oil
High
0.5 oz.
1.515
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H335-May cause respiratory irritation.
  • H371-May cause damage to organs.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : white mineral oil, terphenyls, polyphenyls, polybutane, hydrogenated terphenyls

SDS
Documents

Documents

Certificates
Special Offers

Special Offers