Learn More
Epredia™ Resolve™ Immersion Oil
Eliminate toxicity and disposal problems with the Epredia™ Resolve™ Immersion Oil.
Brand: Epredia™ M4004
Code : A0
Additional Details : CAS Number : 9003-29-6 Weight : 0.01000kg
Description
- Manufactured using a proprietary process that removes interfering substances and will not damage valuable optics or irreplaceable slide specimens
- Packaged conveniently in four sizes
- Available in both low and high viscosity
- Non-drying and non-hardening
- Refractive index of 1.5150
Specifications
Immersion Oil | |
High | |
4 oz. | |
1.515 |
Safety and Handling
- Warning
Hazard Category
- Specific target organ toxicity Category 3
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H335-May cause respiratory irritation.
- H371-May cause damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P271-Use only outdoors or in a well-ventilated area.
- P314-Get medical advice/attention if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P405-Store locked up.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : white mineral oil, polyphenyls, quarterand higher, partially hydrgoenated, hydrogenated terphenyls,