Learn More
Epredia™ Signature Series™ Pen-Fix™
Achieve fixation and dehydration of specimens simultaneously with Epredia™ Signature Series™ Pen-Fix™, a uniquely buffered alcoholic formalin fixative.
£85.95
Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10148438
View Documents
Epredia™
6101
4 x 1 gal.
£85.95
Pack of 4
Description
Description
  • Excellent frozen section fixative
  • Performs well with fatty or thick tissue sections
  • Penetration rate three times faster than 10% Neutral Buffered Formalin
  • Can be used on all open or enclosed tissue processors and in the grossing room
  • Recommended for use in the second and/or third stations of the processor as a secondary fixative