Learn More
Epredia™ Richard-Allan Scientific™ Steiner-Steiner Silver, Silver Nitrate #2 Solution
Determine the presence of spirochetes and nonfilamentous bacteria with Epredia™ Richard-Allan Scientific™ Steiner-Steiner Silver.
Brand:  Epredia™ 87009
 View more versions of this product
Product Code. 10485709
  • £270.00 / Each
Explore more special offers
Description
Description
  • Kit does not contain uranyl nitrate
  • Background is stained yellow to brown
  • Bacteria, Donovan bodies and fungi all stain black
  • Kit formulation and detailed procedures provide consistent and reproducible results
Specifications
Specifications
Steiner-Steiner Silver Kit