Learn More
Epredia™ Richard-Allan Scientific™ Steiner-Steiner Silver, Silver Nitrate #2 Solution
Determine the presence of spirochetes and nonfilamentous bacteria with Epredia™ Richard-Allan Scientific™ Steiner-Steiner Silver.
Brand: Epredia™ 88035
This item is not returnable.
View return policy
Description
- Kit does not contain uranyl nitrate
- Background is stained yellow to brown
- Bacteria, Donovan bodies and fungi all stain black
- Kit formulation and detailed procedures provide consistent and reproducible results
Specifications
Hydroquinone | |
6 Capsules |