missing translation for 'onlineSavingsMsg'
Learn More

BINDER™ Right Access Port (50 to 125 mm)

For Use With: FED 115, FED 260, FED 720

Brand:  BINDER™ 8012-1670

Additional Details : Weight : 0.00100kg

Product Code. 17131391

  • £849.00 / Each
Estimated Shipment: 26-11-2024
to see stock.
Add to basket

Description

Description

  • For use on right
  • Includes silicone plug
  • For use with a wide range of BINDER™ incubators and chambers
Specifications

Specifications

Access port with silicone plug - 100 mm, right
Access Port
100 mm
BINDER Chambers FED 115, FED 260, FED 720
Silicone plug
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers