missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Riluzole hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
25 mg
Description
Inhibits glutamate release from presynaptic terminals and increases Na+-dependent glutamate uptake in rat cortical synaptosomes. Blocks voltage-dependent Na+ channels and inhibits GABA uptake by striatal synaptosomes. Also activates TRPC5 channels. Psychotropic agent with anticonvulsant, hypnotic, anxiolytic, anti-ischemic, anesthetic and neuroprotective properties.
Specifications
Specifications
CAS | 850608-87-6 |
Molecular Formula | C8H6ClF3N2OS |
Quantity | 25 mg |
Synonym | riluzole hydrochloride, prestwick_644, 2-amino-6-trifluoromethoxybenzothiazole hydrochloride, 2-amino-6-trifluoromethoxy-benzothiazole hydrochloride, 2-benzothiazolamine,6-trifluoromethoxy-,monohydrochloride, 2-benzothiazolamine, 6-trifluoromethoxy-, monohydrochloride, 6-trifluoromethoxy-1,3-benzothiazol-2-amine hydrochloride |
InChI Key | QEAOELIJQRYJJS-UHFFFAOYSA-N |
SMILES | C1=CC2=C(C=C1OC(F)(F)F)SC(=N2)N.Cl |
IUPAC Name | 6-(trifluoromethoxy)-1,3-benzothiazol-2-amine;hydrochloride |
Molecular Weight (g/mol) | 270.654 |
PubChem CID | 6419992 |
Formula Weight | 270.66 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Riluzole hydrochloride, Tocris Bioscience™ >
Spot an opportunity for improvement?Share a Content Correction