missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Ring finger protein 214 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30606
This item is not returnable.
View return policy
Description
Ring finger protein 214 Polyclonal antibody specifically detects Ring finger protein 214 in Human samples. It is validated for Immunoprecipitation,Western BlotSpecifications
Ring finger protein 214 | |
Polyclonal | |
Tris-citrate/phosphate pH 7 -8. | |
Rabbit | |
Affinity purified | |
RUO | |
257160 | |
Store at 4°C. Do not freeze. | |
IgG |
Immunoprecipitation, Western Blot | |
Unconjugated | |
DKFZp547C195, ring finger protein 214 | |
Between 1 and 50 | |
100 μg | |
Primary | |
Human | |
Purified |