missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Ring Sets for Precision™ Steaming Water Baths

Precision Ring Sets are for use with steaming water baths.

Brand:  Thermo Scientific™ 3166186

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 16620583

  • £112.72 / 1 set
Estimated Shipment: 25-09-2024
to see stock.
Add to basket

Description

Description

Thermo Scientific™ Precision Ring Sets are for use with steaming water baths.

  • Stainless steel
  • Nest smoothly and become progressively larger as rings are removed
  • Three ring sizes available: 10 cm (4in.), 12.5cm (5in.), and 15cm (6in.)
Specifications

Specifications

Streaming Baths
Ring Set
10 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers