missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ Ringers 085 R-Flex Impact Gloves

Product Code. 17318172
Click to view available options
Size:
10
11
12
3X-Large
7
8
9
This item is not returnable. View return policy
This item is not returnable. View return policy

These Ringers 085 R-Flex Impact Gloves provide innovative impact, cut, and cold protection combined with superior dexterity for ultimate comfort in a variety of end uses.

  • The Ringers 085 R-Flex Impact glove with Nitrile Half Dip and Fleece Lining combines protection against impact hazards and the highest levels of cut resistance, with added warmth for application in cold environments.
  • A breathable knit shell developed with Ringers' F3 Technology™ and patented TPR design for superior dexterity and comfort.
  • The sandy finish palm ensures secure grip on wet and dry surfaces, and touchscreen compatible index, middle and thumb tips allow technology interaction.
TRUSTED_SUSTAINABILITY

Specifications

Applications Industrial
Coating Material Nitrile
Color Multicolored
Hand Specific
Latex Free Yes
Size 7
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'