missing translation for 'onlineSavingsMsg'
Learn More

Ritter Plastic™ Riplate™ 96-Well, Tip Comb for Magnetic Well - U-Shaped-Bottom Microplate

Color:Clear; Lid:Without Lid

Brand:  Ritter Plastic™ 43001-0503

Product Code. 15707938

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • High resistance towards commonly used chemicals, solvents and alcohols
  • High stability also during centrifugation
  • Newly developed cylindrical wells enable a good bioturbation of samples
  • Slightly raised rims reduce the risk of cross-contamination
  • Alphanumeric code allows the registration and identification of individual samples even in confined spaces
  • Especially developed for magnetic separation technology
  • Manufactured from high-purity polypropylene
  • Available in 0.2 mL or 2.0 mL well capacities
Specifications

Specifications

Without Lid
Magnetic 96-well plates
Cylindrical
Clear
96
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers