missing translation for 'onlineSavingsMsg'
Learn More

Ritter Plastic™ Riplate™ 96-Well, Resistant - for Magnetic Separation, V-Shaped-Bottom, 2.0 mL Microplate

Color:Clear

Brand:  Ritter Plastic™ 43001-0502

Product Code. 16397215

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • High resistance towards commonly used chemicals, solvents and alcohols
  • High stability also during centrifugation
  • Newly developed cylindrical wells enable a good bioturbation of samples
  • Slightly raised rims reduce the risk of cross-contamination
  • Alphanumeric code allows the registration and identification of individual samples even in confined spaces
  • Especially developed for magnetic separation technology
  • Manufactured from high-purity polypropylene
  • Available in 0.2 mL or 2.0 mL well capacities
Specifications

Specifications

Magnetic
Clear
KingFisher™ 96 Flex Magnetic Particle Processors, Quagen Bio Sprint 96
2.0 mL
Resistant, for Magnetic Separation
Cleaning, mixing, centrifugation, preparation
Raised rims
60/Pk., 600/Cs.
Well Plate
Virgin Polypropylene
96
Conical Bottom (V-well)
60/Pk.
Cylindrical
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers