missing translation for 'onlineSavingsMsg'
Learn More
Risperidone, 99%, Thermo Scientific Chemicals Product Code.: 15592363

Risperidone, 99%, Thermo Scientific Chemicals

Product Code. 15592363
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15592363

Brand: Thermo Scientific Acros 459390010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106266-06-2
Molecular Formula C23H27FN4O2
Molecular Weight (g/mol) 410.49
InChI Key RAPZEAPATHNIPO-UHFFFAOYSA-N
Synonym risperidone, risperidal, risperdal, risperdal consta, rispolept, risperin, rispolin, sequinan, risperidona, risperidonum
PubChem CID 5073
ChEBI CHEBI:8871
IUPAC Name 3-[2-[4-(6-fluoro-1,2-benzoxazol-3-yl)piperidin-1-yl]ethyl]-2-methyl-6,7,8,9-tetrahydropyrido[1,2-a]pyrimidin-4-one
SMILES CC1=C(C(=O)N2CCCCC2=N1)CCN3CCC(CC3)C4=NOC5=C4C=CC(=C5)F

Specifications

Melting Point 172.0°C
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 410.49
Percent Purity 99%
Chemical Name or Material Risperidone

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0401622 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number459390050,459390010
Lot NumberA0440283 Certificate TypeCertificate of Analysis Date28/03/2022 Catalog Number459390050,459390010
Lot NumberA0422494 Certificate TypeCertificate of Analysis Date21/09/2020 Catalog Number
Lot NumberA0418818 Certificate TypeCertificate of Analysis Date05/05/2020 Catalog Number459390050,459390010
compliance-icons
Product Identifier
  • Risperidone
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Risperidone, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.