missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Risperidone, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Risperidone is an atypical antipsychotic agent that displays 5-HT2A receptor antagonism. Also displays high affinity at D2 receptors (Ki values are 0.4 and 3.13 nM for 5-HT2A and D2 receptors respectively).
Specifications
Quantity | 10 mg |
Formula Weight | 410.48 |
Percent Purity | >99% |
Chemical Name or Material | Risperidone |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'