missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals RITA, p53 activator III

Product Code. 15446869
Click to view available options
Quantity:
1 mg
10 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

A cell-permeable, p53-targeting, tricyclic thiophene derivative

A cell-permeable, p53-targeting, tricyclic thiophene derivative

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 213261-59-7
Molecular Formula C14H12O3S2, C14H12O3S2
Synonym 5,5′-(2,5-Furandiyl)bis-2-thiophenemethanol; NSC 652287

Specifications

Quantity 10 mg
Content And Storage 2°C to 8°C
For Use With (Application) A cell-permeable, p53-targeting, tricyclic thiophene derivative
Formula Weight 292.40g/mol
Sensitivity Light Sensitive
Product Type Signal Transduction Reagent
Form Crystalline solid
InChI Key KZENBFUSKMWCJF-UHFFFAOYSA-N
SMILES OCC1=CC=C(S1)C1=CC=C(O1)C1=CC=C(CO)S1
IUPAC Name (5-{5-[5-(hydroxymethyl)thiophen-2-yl]furan-2-yl}thiophen-2-yl)methanol
Molecular Weight (g/mol) 292.37
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'