missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Rite-On Frosted Thick Slide
Shop All Epredia Products
Click to view available options
Corner Style:
90°
Frosted:
White Epoxy Ink Frosted End, 1 Side
Thickness (Metric):
1 mm
Length (Metric):
75 mm
Unit Size:
Pack of 3600
Description
All finished slides are sealed with a desiccant in an air tight, lint-free, hermetically sealed Moistur-Barrier™ package, keeping slides dry during shipping and storage. Slides work well in hematology, histology, cytology, microbiology and urinalysis; as well as other laboratory and research applications.
- Smooth marking area for effortless writing, automated printing, or printed labels
- Each case features an inner barrier-bag sealed with desiccant packs to protect from moisture contamination
- Slide boxes convert into convenient slide storage files
- Reduced iron content for low self-fluorescence
- Manufactured from pure, water-white glass
- Precleaned for immediate use
- Part No. 3032-002 is used for fluorescent antibody examination
Specifications
Specifications
| Length (Metric) | 75 mm |
| Width (Metric) | 25 mm |
| Edges | Ground |
| Frosted | White Epoxy Ink Frosted End, 1 Side |
| Material | Glass |
| Precleaned | Yes |
| Thickness (Metric) | 1 mm |
| Type | Microscope Slide |
| Color | White |
| Corner Style | 90° |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction