missing translation for 'onlineSavingsMsg'
Learn More

Rivaroxaban, 98%

Product Code. 15527739 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
missing translation for 'unitSize'
1g
250mg
This item is not returnable. View return policy

Product Code. 15527739

missing translation for 'mfr': Thermo Scientific Acros 461602500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 366789-02-8 | C19H18ClN3O5S | 435.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 366789-02-8
Molecular Formula C19H18ClN3O5S
Molecular Weight (g/mol) 435.88
MDL Number MFCD11974010
InChI Key KGFYHTZWPPHNLQ-UHFFFAOYNA-N
Synonym rivaroxaban, xarelto, s-5-chloro-n-2-oxo-3-4-3-oxomorpholino phenyl oxazolidin-5-yl methyl thiophene-2-carboxamide, unii-9ndf7jz4m3, xarelto tn, 9ndf7jz4m3, 5-chloro-n-5s-2-oxo-3-4-3-oxomorpholin-4-yl phenyl-1,3-oxazolidin-5-yl methyl thiophene-2-carboxamide, 2-thiophenecarboxamide, 5-chloro-n-5s-2-oxo-3-4-3-oxo-4-morpholinyl phenyl-5-oxazolidinyl methyl, 5-chloro-n-5s-2-oxo-3-4-3-oxo-4-morpholinyl phenyl-5-oxazolidinyl methyl-2-thiophenecarboxamide
PubChem CID 9875401
ChEBI CHEBI:68579
IUPAC Name 5-chloro-N-[[(5S)-2-oxo-3-[4-(3-oxomorpholin-4-yl)phenyl]-1,3-oxazolidin-5-yl]methyl]thiophene-2-carboxamide
SMILES ClC1=CC=C(S1)C(=O)NCC1CN(C(=O)O1)C1=CC=C(C=C1)N1CCOCC1=O

Specifications

Melting Point 228.0°C to 229.0°C
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 250 mg
Formula Weight 435.88
Percent Purity 98%
Chemical Name or Material Rivaroxaban
compliance-icons compliance-icons
Product Identifier
  • Rivaroxaban
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P273-Avoid release to the environment.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P391-Collect spillage.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.