missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RJR 2403 oxalate, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
A neuronal nicotinic receptor agonist, showing high selectivity for the α4β2 subtype (Ki values are 26 and 36000 nM for α4β2 and α7 receptors respectively). Active in vivo.
Specifications
Specifications
Quantity | 50 mg |
Formula Weight | 252.27 |
Percent Purity | >99% |
Chemical Name or Material | RJR 2403 oxalate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction