missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RN 1 dihydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent lysine specific demethylase 1 (LSD1) inhibitor (IC50 = 70 nM). Exhibits selectivity for LSD1 over MAO-A and MAO-B (IC50 values are 0.51 and 2.79 μM for MAO-A and MAO-B, respectively). Impairs long-term, but not short-term memory in mice. Brain penetrant following systemic administration.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 452.42 |
Chemical Name or Material | RN 1 dihydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'