missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNA Control 250 with Manual

Product Code. 10400725
Click to view available options
Quantity:
2 x 20 μL, with Manual
2 x 20μL
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10400725

Brand: Invitrogen™ AM7155

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

RNA Control and THE RNA Storage Solution

The Invitrogen RNA Control 250 provides a precise, accurate, and reproducible RNA concentration reference for use in conjunction with NanoDrop™ Spectrophotometers.

The Invitrogen RNA Control 250 provides a precise, accurate, and reproducible RNA concentration reference for use in conjunction with NanoDrop™ Spectrophotometers. It is provided in two tubes containing 20 μL each. RNA Control 250 consists of a single, ultra-pure, 2000nt RNA transcript of a precisely defined sequence, molecular weight, and concentration. It is packaged in special non-stick tubes and includes the Invitrogen THE RNA Storage Solution. The RNA Control 250 will maintain ± 5% of stated concentration and >85% RNA integrity through 5 freeze/thaw cycles (10 duplicate measurements).

• A stable, highly pure transcript at a precisely known concentration
• Used to verify the accuracy of NanoDrop Spectrophotometer measurements
• Provided with THE RNA Storage Solution for blank measurements
• Enough for 20 measurements

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage RNA Control and THE RNA Storage Solution should be stored at –20°C
Format Tube
Label or Dye Unconjugated
Product Line Ambion
Quantity 2 x 20μL
Shipping Condition Dry Ice
Volume 2 x 20 μL

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.