Learn More
Invitrogen™ RNA Control 250 with Manual
Description
The Invitrogen RNA Control 250 provides a precise, accurate, and reproducible RNA concentration reference for use in conjunction with NanoDrop™ Spectrophotometers. It is provided in two tubes containing 20 μL each. RNA Control 250 consists of a single, ultra-pure, 2000nt RNA transcript of a precisely defined sequence, molecular weight, and concentration. It is packaged in special non-stick tubes and includes the Invitrogen THE RNA Storage Solution. The RNA Control 250 will maintain ± 5% of stated concentration and >85% RNA integrity through 5 freeze/thaw cycles (10 duplicate measurements).
• A stable, highly pure transcript at a precisely known concentration
• Used to verify the accuracy of NanoDrop Spectrophotometer measurements
• Provided with THE RNA Storage Solution for blank measurements
• Enough for 20 measurements
Specifications
Specifications
| Content And Storage | RNA Control and THE RNA Storage Solution should be stored at –20°C |
| Format | Tube |
| Label or Dye | Unconjugated |
| Product Line | Ambion |
| Quantity | 2 x 20 μL, with Manual |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.