Learn More
Cytiva RNA Homopolymers
Template primers
Brand:  Cytiva 27-4110-02
Product Code. 11585115
  • £973.00 / 500mg
Explore more special offers
Description
Description
These products are dispensed by weight of dry material, which includes polymer, residual salt, and residual water; content of polymer may vary from lot to lot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RNA Homopolymer
-20°C
Poly(rA)
500 mg