missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ RNA UltraSense™ One-Step Quantitative RT-PCR System

Product Code. 10616743 Shop All Applied Biosystems Products
Click to view available options
Quantity:
100 rxns
Unit Size:
100 reactions
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10616743

Brand: Applied Biosystems™ 11732927

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

RNA UltraSense™ One-Step Quantitative RT-PCR System

The RNA UltraSense™ One-Step Quantitative RT-PCR System is specially designed for amplification and real-time quantification of ultra-low abundance transcripts and RNA viruses. The optimized, ultra-concentrated system combines SuperScript™ III RT and Platinum™ Taq DNA Polymerase to provide greater priming specificity, higher product yields, and detection over a broad dynamic range. The high concentration formulation provides great flexibility with low concentration / high volume RNA samples, and improves performance with targets rich in secondary structure. As little as 25 pg/μl total RNA can be detected with a PCR efficiency of 97% (Figure 1). The RNA UltraSense™ System features:

  • SuperScript™ III RT for higher temperature cDNA synthesis (up to 60°C) for greater success with RNA secondary structure
  • Platinum™ Taq DNA Polymerase with hot-start technology for improved specificity
  • Optimized performance with either LUX™ Fluorogenic Primers or dual-labeled fluorogenic probes
  • Proprietary, ultra-concentrated buffer for addition of large sample volumes (up to 70%) and success with RNA secondary structure
  • Validated performance on multiple real-time instrument platforms
  • One-step format for speed, convenience, and less reaction-to-reaction variability

    Applications:
    Real-time quantitative amplification of RNA, including:
  • RNA viruses
  • Low-abundance RNA transcripts
  • Difficult-to-amplify RNA targets
  • High-value RNA detection and research
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage The RNA UltraSense™ One-Step Quantitative RT-PCR System is supplied with the following components: 250 μl RNA UltraSense™ Enzyme Mix (includes SuperScript™ III RT, Platinum™ Taq DNA Polymerase, and RNaseOUT™ Ribonuclease Inhibitor); 2 x 1 ml RNA UltraSense™ 5X Reaction Mix; 300 μl 20X Bovine Serum Albumin (BSA), Ultrapure Molecular Biology Grade (5 mg/ml); 1 ml 50-mM magnesium sulfate (MgSO4 ); 100 μl ROX Reference Dye. Sufficient reagents are provided for 100 reactions. Store components at -20°C. Stability can be extended by storing at -80°C. ROX Reference Dye must be stored in the dark.
    Detection Method Primer-probe
    GC-Rich PCR Performance High
    PCR Method 1-step RT-qPCR
    Polymerase DNA Polymerase
    Reaction Speed Standard
    Product Line Platinum, SuperScript, UltraSense
    Product Type One-Step Quantitative RT-PCR System
    Quantity 100 rxns
    Shipping Condition Dry Ice
    Sufficient For 100 Reactions
    No. of Reactions 100 Reactions
    Sample Type RNA
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.