Learn More
Thermo Scientific™ RNase AWAY™ Surface Decontaminant
Eliminate RNase and DNA from laboratory surfaces. This product reduces dependency on carcinogenic DEPC treatments and saves time spent baking glassware.
Brand: Thermo Scientific™ 7002PK
194.22 GBP valid until 2024-12-31
Use promo code "23291" to get your promotional price.
Additional Details : Weight : 0.10000kg
Description
Eliminate RNase and DNA from laboratory surfaces. Thermo Scientific™ RNase AWAY™ is ideal for decontaminating apparatus, bench-tops, glassware and plasticware. It reduces dependency on carcinogenic DEPC treatments and saves time needed to bake glassware.
- Use on pipettes, gel boxes, or RNA or DNA prep areas
- Leaves no residue to interfere with gel polymerization or staining
- Chemically stable and nonabrasive
- Contains no strong acids