Learn More
Macherey-Nagel™ RNaseA
Intended primarily for use with NucleoSpin™ and NucleoBond™ kits. Macherey-Nagel™ RNase A is a freeze-dried enzyme sourced from bovine pancreas. It has RNase activity of 120 U/mg. Available in 50mg and 100mg packs.
£108.00 - £197.00
Specifications
Color | White |
---|---|
For Use With (Equipment) | NucleoSpin™ and NucleoBond™ kits |
Storage Buffer | A1 / S1 / S1-EF / RES / RESEF / RES-BAC |
Content And Storage | Store at room temperature (18–25°C) for at least 1 year |
Source | Bovine pancreas |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10174711
|
Macherey-Nagel™
740505.50 |
50 mg |
£108.00
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10546881
|
Macherey-Nagel™
740505 |
100 mg |
£197.00
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ribonuclease (RNase) is an enzyme that catalyzes the degradation of RNA into smaller components. RNase A is one of the hardiest enzymes in common laboratory usage. It is specific for single-stranded RNAs, cleaving the 3'-end of unpaired C and U residues, and ultimately forming a 3'-phosphorylated product via a 2',3'-cyclic monophosphate intermediate. It does not require any cofactors for its activity.
- CAS: 9001-99-4
- For research purposes only
- For use, dissolve in water or resuspension buffer
- Dissolved in water, it can be stored at 4°C for up to 3 months (in smaller aliquots, at −20°C, for longer)
- Dissolved in resuspension buffer A1 / S1 / S1-EF / RES / RESEF / RES-BAC, it can be stored at 4°C for up to 6 months
Specifications
White | |
A1 / S1 / S1-EF / RES / RESEF / RES-BAC |
|
Bovine pancreas | |
Freeze-dried (Lyophilized) |
NucleoSpin™ and NucleoBond™ kits | |
Store at room temperature (18–25°C) for at least 1 year | |
Ribonuclease A |
Safety and Handling
- Danger
Hazard Category
- Respiratory sensitiser Category 1
Hazard Statement
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .