Learn More
Applied Biosystems™ RNaseP Verification Plate, 96-well
The 96-Well RNaseP Verification Plate is used to verify your real-time PCR instrument with Standard 96-Well Block
Brand:  Applied Biosystems™ 4432593
Product Code. 15798669
  • £890.00 / Each
Explore more special offers
Description
Description
  • The 96-Well RNaseP Verification Plate is used to verify your real-time PCR instrument with Standard 96-Well Block.
  • Confirm the performance of your instrument by running the RNaseP assay. Please see your instrument user guide for full instructions on use.
  • The 96-Well RNaseP Verification Plate is preloaded with the necessary reagents to detect and quantitate genomic copies of the human RNase P gene, a single-copy gene encoding the RNA moiety of the RNase P enzyme.
  • Each well contains preloaded reaction mix (1X TaqMan™ Universal PCR Master Mix, RNase P primers, and FAM™ dye-labeled probe) and template.
  • The verification plate must demonstrate the ability to distinguish between 5,000 and 10,000 genomic equivalents with a 99.7% confidence level for a subsequent sample run in a single well.
  • To ensure accurate results, it is recommended that the performance of your instrument be verified every six months.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Store at –20 ± 5°C.
Proprietary Mix
ViiA 7 Dx, QuantStudio Dx, QuantStudio 5 Dx, QuantStudio 7 Pro Dx
Instrument Verification Plate
Dry Ice
1 x 96-well plate