missing translation for 'onlineSavingsMsg'
Learn More

Ro 20-1724, Tocris Bioscience™

Product Code. 30001086
Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

PDE4 inhibitor

Ro 20-1724 is a widely used inhibitor of cyclic nucleotide phosphodiesterase, selective for PDE4 (IC50 = 2.0 μM).

Specifications

CAS 29925-17-5
Molecular Formula C15H22N2O3
Quantity 50 mg
Synonym 4-3-butoxy-4-methoxybenzyl-2-imidazolidinone, 4-3-butoxy-4-methoxybenzyl imidazolidin-2-one, 4-3-butoxy-4-methoxyphenyl methyl imidazolidin-2-one, chembl18701, 2-imidazolidinone, 4-3-butoxy-4-methoxyphenyl methyl, 2-imidazolidinone, 4-3-butoxy-4-methoxyphenyl methyl-, +-, 4-3-butoxy-4-methoxyphenyl methyl-2-imidazolidone, keratan sulfate i, keratan sulphate i, naphthoquinone analogs
InChI Key PDMUULPVBYQBBK-UHFFFAOYSA-N
SMILES CCCCOC1=C(C=CC(=C1)CC2CNC(=O)N2)OC
IUPAC Name 4-[(3-butoxy-4-methoxyphenyl)methyl]imidazolidin-2-one
Molecular Weight (g/mol) 278.352
PubChem CID 5087
Formula Weight 278.35
Percent Purity >99%
Chemical Name or Material Ro 20-1724
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'