missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ro 32-0432 hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
1 mg
Description
Selective cell-permeable protein kinase C inhibitor. Displays slight selectivity for conventional PKC isoforms over Ca2+ and atypical PKC isoforms; binding affinities for rat isoforms are 9, 28, 31, 37 and 108 nM for PKC's α, βΙ, βΙΙ, γ and ε respectively. Orally available and prevents T cell chronic inflammation in vivo.
Specifications
CAS | 1781828-85-0 |
Molecular Formula | C28H29ClN4O2 |
Quantity | 1 mg |
Synonym | ro 32-0432 hydrochloride |
InChI Key | HSPRASOZRZDELU-UNTBIKODSA-N |
SMILES | CN1C=C(C2=CC=CC=C21)C3=C(C(=O)NC3=O)C4=C5CC(CCN5C6=CC=CC=C64)CN(C)C.Cl |
IUPAC Name | 3-[(8R)-8-[(dimethylamino)methyl]-6,7,8,9-tetrahydropyrido[1,2-a]indol-10-yl]-4-(1-methylindol-3-yl)pyrrole-2,5-dione;hydrochloride |
Molecular Weight (g/mol) | 489.016 |
PubChem CID | 124081018 |
Formula Weight | 489.01 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'