missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Ro 48-8071 fumarate

2,3-Oxidosqualene cyclase (OSC) inhibitor; blocks cholesterol synthesis

£106.00 - £353.00

Specifications

Purity 99%
Molecular Formula (4-Bromophenyl)[2-fluoro-4-[[6-(methyl-2-propen-1-ylamino)hexyl]oxy]phenyl]methanone
CAS 189197-69-1
Content And Storage 4°C
Product Type Ro 48-8071 fumarate
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18712418
View Documents Promotion Details
Tocris Bioscience™
5389/50
50 mg
£353.00
50mg
Please to purchase this item. Need a web account? Register with us today!
18797548
View Documents
Tocris Bioscience™
5389/10
10 mg
£106.00
10mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

2,3-Oxidosqualene cyclase (OSC) inhibitor (IC50 = 6.5 nM); blocks cholesterol synthesis in HepG2 cells. Reduces viability of breast cancer cell lines. Also inhibits Ebola virus (EBOV) cell entry (IC50 = 1.74 μM).
Specifications

Specifications

99%
189197-69-1
Ro 48-8071 fumarate
Ro 48-8071 fumarate
(4-Bromophenyl)[2-fluoro-4-[[6-(methyl-2-propen-1-ylamino)hexyl]oxy]phenyl]methanone
4°C
M.W. (Observed): 564.44
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only