Learn More
RO-9187, MedChemExpress
MedChemExpress RO-9187 is a potent inhibitor of HCV virus replication with an IC50 of 171 nM.

£240.79 - £960.74
Specifications
Chemical Name or Material | RO-9187 |
---|---|
CAS | 876708-03-1 |
Color | Light Yellow |
Purity Grade Notes | Research |
Molecular Formula | C9H12N6O5 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16456947
![]() |
Medchem Express
HY-10870 |
5 mg |
£240.79
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16444228
![]() |
Medchem Express
HY-10870 |
10 mg |
£408.98
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16440567
![]() |
Medchem Express
HY-10870 |
50 mg |
£960.74
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RO-9187 is a potent inhibitor of HCV virus replication with an IC50 of 171 nM.Specifications
RO-9187 | |
Light Yellow | |
C9H12N6O5 | |
O=C1N=C(N)C=CN1[C@H]2[C@@H](O)[C@H](O)[C@](CO)(N=[N+]=[N-])O2 | |
284.23 | |
Research | |
COVID-19-anti-virus |
876708-03-1 | |
Research | |
DMSO : 100 mg/mL (351.83 mM; Need ultrasonic) ∣H2O : 7.14 mg/mL (25.12 mM; Need ultrasonic) | |
284.23 | |
98.0% | |
Solid |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.