Learn More
BRAND™ Robotic Tips, BIO-CERT™ Certified Quality
Shop All Brand Products
Click to view available options
Volume (Metric):
1 to 50 μL
10 to 200 μL
10 to 300 μL
40 to 1000 μL
Unit Size:
Pack of 960
Description
- The raw materials are free from DiHEMDA und Oleamid. Tips are manufactured at BRAND under monitored cleanroom conditions and are automatically packaged. No parting agents or demolding aids are used. All racked tips and filter tips up to 1000 μL are free of DNA (< 40 fg), RNase (< 8.6 fg), endotoxins (< 1 pg) and ATP (< 1 fg).
- For use with pipetting robots, tips are required that are matched to the robot's mode of operation in terms of geometry and surface properties. BRAND robotic pipette tips and robotic filter tips meet all requirements for use with the Liquid Handling Station pipetting robot.
Specifications
Specifications
| Volume (Metric) | 10 to 300 μL |
| Filtered | Filtered |
| Sterility | Non-sterile |
| Packaging | TipRack (960/Pk.) |
| Product Type | Robotic Tips |
| Material | Polypropylene |