Learn More
BRAND™ Robotic Tips, BIO-CERT™ Certified Quality
Pipette tips offer reliable quality for precise and demanding analyses with the pipetting robot Liquid Handling Station.
Brand:  BRAND™ 734152
64.79 GBP valid until 2025-04-13
Use promo code "24122" to get your promotional price.
 View more versions of this product
Product Code. 17556696
  • £77.75 / Pack of 960
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The raw materials are free from DiHEMDA und Oleamid. Tips are manufactured at BRAND under monitored cleanroom conditions and are automatically packaged. No parting agents or demolding aids are used. All racked tips and filter tips up to 1000 μL are free of DNA (< 40 fg), RNase (< 8.6 fg), endotoxins (< 1 pg) and ATP (< 1 fg).
  • For use with pipetting robots, tips are required that are matched to the robot's mode of operation in terms of geometry and surface properties. BRAND robotic pipette tips and robotic filter tips meet all requirements for use with the Liquid Handling Station pipetting robot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
40 to 1000 μL
Non-sterile
Robotic Tips
Filtered
TipRack (960/Pk.)
Polypropylene
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BRAND™ Robotic Tips, BIO-CERT™ Certified Quality > Volume: 40 to 1000 μL