missing translation for 'onlineSavingsMsg'
Learn More

Rolitetracycline, MedChemExpress

Product Code. p-200101469
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16489967

Brand: Medchem Express HY18257

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Rolitetracycline, a derivative of tetracycline, is a broad-spectrum antibiotic. Rolitetracyclin has a role as a protein synthesis inhibitor, an antiprotozoal drug and a prodrug.

Rolitetracycline, a derivative of tetracycline, is a broad-spectrum antibiotic. Rolitetracyclin has a role as a protein synthesis inhibitor, an antiprotozoal drug and a prodrug.

Specifications

Chemical Name or Material Rolitetracycline
CAS 751-97-3
Color Light Brown
Purity Grade Notes Research
Molecular Formula C27H33N3O8
Quantity 5 mg
Solubility Information H2O : 10 mg/mL (18.95 mM; Need ultrasonic and warming) ∣Ethanol : < 1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble) ∣DMSO : < 1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble or slightly soluble)
SMILES O=C(C(C1=O)=C(O)[C@@H](N(C)C)[C@]2([H])C[C@]3([H])[C@](C)(O)C4=C(C(C3=C(O)[C@@]21O)=O)C(O)=CC=C4)NCN5CCCC5
Molecular Weight (g/mol) 527.57
Formula Weight 527.57
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.