missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Roll Form European Standard Pipemarkers with Die-cut Arrowheads - Air
European Standard Pipemarkers increase safety and aid maintenance.
Brand: Brady™ 228208
Code : 50
Additional Details : Weight : 1.35000kg
Description
- Pipemarkers conform to RAL European standard color coding and for dangerous substances.
- The Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
- Danger and hazard symbols, if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, which saves you having to identify the appropriate symbol.
- Markers are printed with a contrasting border in the same color as the text for improved visibility.
- The design and layout of Brady's roll form markers ensure maximum visibility from all sides.
- Parts supplied with a black arrow printed on white background.
Specifications
European Directives 67/548 and 88/379 | |
Permanent Acrylic | |
Gloss | |
Polyester | |
Instrument Air | |
English | |
505 Pk. | |
-40°C to +110°C | |
0.099 mm | |
33 m x 100 mm x 60 mm | |
33 m | |
100 mm | |
0.31 in. | |
8 mm |
Roll form Pipe Markers with liner, without pictograms - Air - Instrument air | |
White on Blue, Blue | |
Roll | |
Pipe Marker | |
Pipe and valve marking | |
505 Markers/Roll | |
-40°F to +230°F | |
0.0039 in. | |
33 m x 100 mm | |
108.27 ft. | |
3.94 in. | |
2.36 in. | |
60 mm |