missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Roll-Form Linerless Pipe Marker - Water

Brand:  Brady™ 224388

Code : 50

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12784309

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

ROHS Environmental Compliance, EU Directive 2002/95/EC
Acrylic, Permanent, Pressure-sensitive
White, Green
Roll Form Linerless Pipe Marker
Used for linerless Pipemarkers
505/Pk.
0.120 mm
100 mm
8 mm
Roll form linerless Pipe Markers, without pictograms - Water
Silicone
Laminated Polyester
Chilled Water
English
-40°C to +140°C
60 x 100 mm
60.00 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers