missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Roll Form Linerless Pipe Markers, without Pictograms - Water
European Standard Pipemarkers reinforce safety and aid maintenance.
Brand: Brady™ N008662
Code : NEW
Additional Details : Weight : 0.20000kg
Description
- Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
- Danger and hazard symbols, if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, this saves you having to identify the appropriate symbol.
- Markers are printed with a contrasting border in the same color as the text for improved visibility.
- The design and layout of Brady's roll form markers ensure maximum visibility from all sides.
- Parts supplied with a Black arrow printed on white background.
Specifications
Roll form linerless Pipe Markers, without pictograms - Water - Condensate | |
Silicone | |
Roll | |
Pipe Marker | |
Pipe and valve marking | |
220 Markers/Roll | |
-40°F to +284°F | |
0.0047 in. | |
33 m x 100 mm | |
108.27 ft. | |
3.94 in. | |
5.91 in. | |
13.6 mm |
Permanent Acrylic | |
White on Green | |
Polyester | |
Condensate | |
English | |
220 Pk. | |
-40°C to +140°C | |
0.119 mm | |
33 m x 100 mm x 150 mm | |
33 m | |
100 mm | |
150 mm |