missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Roll of hybridisation oven mesh, 5m x 25cm

Brand:  Thermo Scientific™ 222054

Product Code. 12624653

  • £467.02 / Each
Estimated Shipment: 02-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Manufactured using thick walled borosilicate glass designed to ensure safe and easy handling
  • Easy flow of probe solution around and through membranes designed for optimal results
  • Designed to exact safety standards, these bottles have screw caps containing flat PTFE seals designed to ensure leak-resistant hybridisation
  • A full range of bottles to accommodate many sizes of membrane
  • Enables multiple blots to be processed simultaneously in one 35mm diameter bottle without direct overlap
Dimensions: 5m × 25cm
TRUSTED_SUSTAINABILITY
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers