missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Roll Stand with Cutting Edge
The Roll Stand with Cutting provides safe and easy storage and separation of the roll.
£111.00 - £242.00
Product Code | Brand | Width (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Width (Metric) | Price | Quantity & Availability | |||||
17836506
|
asecos™
9908 |
450.00 mm |
£111.00
Each |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
17896486
|
asecos™
9909 |
880.00 mm |
£242.00
Each |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
Description
- Roll stand for easy and safe storage
- Steel Cutting Edge for easy separation
Specifications
Steel | |
Spill Kit Dispenser |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title