Learn More
Invitrogen™ ROR1 Monoclonal Antibody (2H6)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA515539
Description
MA5-15539 targets ROR1 in indirect ELISA, IF and WB applications and shows reactivity with Human samples. The MA5-15539 immunogen is recombinant extracellular fragment of human ROR1 (aa30-406) fused with hIgGFc tag, expressed in HEK293 cells. MA5-15539 detects ROR1 which has a predicted molecular weight of approximately 101kDa.
Receptor-tyrosine-kinase-like orphan receptor 1 (ROR1) is a tumor-associated, surface protein predominantly expressed during embryogenesis, where it is involved in organ morphogenesis, nervous system development, and neural progenitor cell maintenance and survival. Virtually absent from normal pediatric and adult tissues, with the exception of low-level expression in a subset of immature b cell precursors known as hematogones and adipocytes, ROR1 is notably overexpressed, and considered a survival factor, in a number of B lymphoid and epithelial malignancies: including chronic lymphocytic leukemia (CLL), mantle cell lymphoma (MCL), acute lymphoblastic leukemia (ALL), marginal zone lymphoma, lung adenocarcinoma. First identified during PCR-based cloning of a human neuroblastoma cell line in search of tyrosine kinases similar to tropomyosin-receptor-kinase (Trk) neurotropic receptors, ROR1, along with the related receptor tyrosine kinase (RTK) ROR2, was catalogued as an orphan receptor due to the fact its related ligand remained elusive. Wnt-5a has since been suggested as a candidate ligand for ROR1, and ROR1 has been implicated to function as a pseudokinase, promoting proliferation and resistance to apoptosis in cancer cells through interaction with Wnt-5a, and TCL1-co-activation of AKT. ROR1 is expressed as a glycoprotein containing extracellular immunoglobulin (Ig)-like, Frizzled, and Kringle domains, as well as an intracellular region containing a tyrosine kinase domain.
Specifications
| ROR1 | |
| Monoclonal | |
| Conc. Not Determined | |
| ascites with 0.03% sodium azide | |
| Q01973 | |
| ROR1 | |
| Recombinant extracellular fragment of human ROR1 (aa30-406) fused with hIgGFc tag, expressed in HEK293 cells. | |
| RUO | |
| 4919 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 2H6 | |
| Unconjugated | |
| ROR1 | |
| 2810404D04Rik; dJ537F10.1; Inactive tyrosine-protein kinase transmembrane receptor ROR1; MGC99659; mRor1; neurotrophic tyrosine kinase, receptor related 1; neurotrophic tyrosine kinase, receptor-related 1; Ntrkr1; receptor tyrosine kinase like orphan receptor 1; receptor tyrosine kinase-like orphan receptor 1; RGD1559469; ROR1; T-ROR1; tyrosine-protein kinase transmembrane receptor ROR1 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.