Learn More
Description
Rose Bengal is a stain used in the eye drops to identify the eye damage. It finds application in the preparation of foraminifera for microscopic analysis, photosensitizing reactions and as a dye. In synthetic chemistry, it is utilized to convert triplet oxygen to singlet oxygen, which is used in [2+2] cycloadditions with alkenes. Its derivatives have important medical functions and used for the treatment of cancer. It acts as an alternative to phloxine B in Kreyberg's stain for keratin and mucus. As a fluorescent probe, it is used for the determination of NAD binding sites of alcohol dehydrogenase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Soluble in water.
Notes
Incompatible with strong oxidizing agents.

Specifications
Specifications
Melting Point | >300°C |
Quantity | 25 g |
Beilstein | 3645857 |
Sensitivity | Light sensitive |
Merck Index | 14,8262 |
Solubility Information | Soluble in water. |
Formula Weight | 1017.65 |
Chemical Name or Material | Rose Bengal |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10253219 | Certificate of Analysis | 16/12/2024 | A17053.14,A17053.36,A17053.22 |
10251749 | Certificate of Analysis | 30/08/2024 | A17053.14,A17053.36,A17053.22 |
10169143 | Certificate of Analysis | 23/11/2021 | A17053.14,A17053.36,A17053.22 |
10206233 | Certificate of Analysis | 23/11/2021 | A17053.14,A17053.36,A17053.22 |
10195076 | Certificate of Analysis | 23/11/2021 | A17053.14,A17053.36,A17053.22 |
Safety and Handling

- Rose Bengal
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P405-Store locked up.
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
Special Offers
-
Multiple Manufacturers
Chemical Extravaganza
Shop now and save up to 97% on top-quality products for your lab needs.Promo Code: 25908Expires: 2025-03-31
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.