missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rotaflo™ Straight Pattern Plain-side Arms for Burettes
Chemical resistant borosilicate glass with PTFE key and rotary design for fine single drop control
Brand: Quickfit GP10ST/10
This item is not returnable.
View return policy
Description
- No lubrication required - no seizure and no contamination
- Temperature range: -20°C to 200°C
- Vacuum capability to 10 Torr
- Increased barrel diameter for drainage
- Locking ring to prevent accidental removal
- Ergonomic key for good grip and easy operation
Specifications
Replacement Stopcock | |
13 mm | |
13 mm | |
-20°C to 200°C | |
With PTFE Key |
10 mm | |
Borosilicate Glass | |
225 mm | |
GP Rotaflo™ Stopcock |