missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heidolph™ Rotamax 120
Compact model with a maximum load capacity of 2 kg.
Brand: Heidolph™ 544-41200-06
970.65 GBP valid until 2024-12-15
Use promo code "21658" to get your promotional price.
This item is not returnable.
View return policy
Description
- Space-saving and with a load weight of up to 2 kg, this shaker can perfectly master all applications.
- The variably adjustable speed control (20 to 300 rpm) and the orbit of 20 mm guarantee homogeneous distribution. Your applications can be carried out in continuous operation or by means of a timer (max. 120 minutes).
Specifications
230 V | |
UK Plug |