Learn More
Hirschmann Instruments™ rotarus™ PK 10 Peristaltic Pump Heads
Facilitates quick and simple head changing in a single action without tools. Hirschmann Instruments™ rotarus™ Peristaltic Pump Heads were developed for rapid handling.
Brand: Hirschmann Instruments™ 951102020
This item is not returnable.
View return policy
Description
- 3 rolls
- For WS/WT 1.6mm tubes
Specifications
rotarus PK 10 Pump Head | |
1 | |
3 | |
WT 16mm |
Product Content Correction
Product Title
Hirschmann Instruments™ rotarus™ PK 10 Peristaltic Pump Heads >
Spot an opportunity for improvement?Share a Content Correction