Learn More
Gardner Denver Welch™ Rotary Evaporator Connection Kit
Used for chemical duty diaphragm pumps.
Brand: Gardner Denver Welch™ 112575
This item is not returnable.
View return policy
Description
- Easy connection of the pump to common rotary evaporators
- Includes vacuum hose 18/8x5 (1.5m), cooling water, chiller connection hose and clamps
Specifications
Rotary Evaporator Connection Kit | |
Chemical duty diaphragm pumps |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Fisherbrand Vacuum pumps
File size: 5.6 MB
Product Content Correction
Product Title
Gardner Denver Welch™ Rotary Evaporator Connection Kit >
Spot an opportunity for improvement?Share a Content Correction