missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vacuubrand™ Rotary Vane Pump RZ 9
Rotary vane pumps offer high performance in a compact design. Even with the gas ballast open, they enable an ideal ultimate vacuum. You also benefit from extended oil change and maintenance intervals thanks to its optimized lubrication circuit and large usable oil volume.
Brand: Vacuubrand™ 20798142
3054.25 GBP valid until 2024-12-31
Use promo code "21962" to get your promotional price.
This item is not returnable.
View return policy
Description
- Very high flow rates, even at vacuum levels approaching ultimate vacuum
- High water vapor tolerance due to efficient gas ballast
- Ideal ultimate vacuum even with gas ballast
- Vacuum-tight at switch-off; external anti-suckback valve not needed
- Large oil volume allows for long intervals between oil changes
- Ease of maintenance due to telescopic design
Specifications
2 x 10-3 mbar | |
UK-Style, India | |
Rotary Vane Vacuum Pump | |
Drying Chambers, Lyophilization, Fine Vacuum Distillation | |
230 V |
503 x 156 x 240 mm | |
8.9 m3/hr. | |
IP40 | |
1.1 bar |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction