Learn More
Fisherbrand™ Rotary Vane Vacuum Pump
Designed for laboratory applications requiring deep vacuum. Fisherbrand™ Rotary Vane Vacuum Pump is a two-stage direct driven rotary vane pumps that provide a high water vapor tolerance.
Brand:  Fisherbrand™ 302108
 View more versions of this product
Product Code. 12931141
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Two-stage direct driven rotary vane pump
  • Designed for wide range of physical and chemical laboratory applications and continuous running
  • Low weight, small footprint and high pumping speed, increased chemical resistance are advantageous features in research and routine processes
  • Rugged reliable design, combined with low noise level and ease of use
  • Innovative gas ballast valve prevents moisture condensing and high tolerance of water and solvent vapors
  • Intake line is vacuum sealed by and anti suckback valve to protect your application from undesired venting and oil back flow after pump is turned off
  • Vertical oil sight glass allows easy monitoring of oil level and condition
Specifications
Specifications
2 x 10-3 mbar
44.5 x 15.0 x 23.5 cm (17.5 x 5.9 x 9.25 in.)
47.3 lb.
120/143 L/min.
50/60 Hz
120 L/min.
2 x 10-3 mbar
115/230 V 50/60 Hz motor with Schuko UK and US power leads
90 w
21.5 kg
Labovac 10
Rotary Vane Vacuum Pump
Freeze Drying
230 V
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products